kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
Standards
Organic Standards
Organic reference standards, leflunomides, Ph. Eur.
Organic reference standards, leflunomides, Ph. Eur.
:
EDQM
Organic reference standards, leflunomides, Ph. Eur.
:
EDQM
CAS No.:
75706-12-6
Formula:
C₁₂H₉F₃N₂O₂
Description:
Leflunomide