kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
Standards
Organic Standards
Organic reference standards, indometacins, Ph. Eur.
Organic reference standards, indometacins, Ph. Eur.
:
EDQM
Organic reference standards, indometacins, Ph. Eur.
:
EDQM
CAS No.:
53-86-1
Formula:
C₁₉H₁₆ClNO₄
Description:
Indometacin