VWR® B'Wipes 4000, Cleanroom wipes
Supplier: VWR Collection
Certificates
About this item
Polyester, sealed-edge wipe for use in ISO 3 cleanrooms.
- Single-knit, polyester wipe, with sealed edges (150 g/m²)
- Laundered, with sealed edges, for superior cleanliness
- Good mechanical resistance and absorption capacity
- Ideal for cleaning and disinfecting surfaces in critical environments