Anti-IgG Donkey Polyclonal Antibody (Biotin)
Catalog # 30-8080-M001
Supplier: Tonbo Biosciences
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Donkey
- ImmunoChemistry:Yes
- Reactivity:Goat
- Epitope:Heavy and light chain
- Cross adsorption:Yes
- Storage temperature:Store at 2-8 °C
- Pk:1 mg
Specifications
About this item
Donkey anti-Goat IgG (H&L) - Affinity Pure, Biotin Conjugate, min x w/human, mouse, rabbit or rat IgG
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and light chain
Host: Donkey
Isotype:
Reactivity: Goat