Order Entry
Norway
ContactUsLinkComponent
Anti-IgM Goat Polyclonal Antibody (Biotin)
Anti-IgM Goat Polyclonal Antibody (Biotin)
Catalog # 30-8064-M002
Supplier:  Tonbo Biosciences
Anti-IgM Goat Polyclonal Antibody (Biotin)
Catalog # 30-8064-M002
Supplier:  Tonbo Biosciences

Specifications

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin M
  • Antigen symbol:
    IgM
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Mouse
  • Epitope:
    Heavy and light chain
  • Storage temperature:
    Store at 2-8 °C
  • Pk:
    2 mg

Specifications

About this item

Goat anti-Mouse IgM (H&L) - Affinity Pure, Biotin Conjugate

Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and light chain
Host: Goat
Isotype:
Reactivity: Mouse