Anti-IgA Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 35-8023-M001
Supplier: Tonbo Biosciences
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin A alpha
- Antigen symbol:IgA
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Epitope:α chain
- Cross adsorption:Yes
- Storage temperature:Store at 2-8 °C
- Pk:1 mg
Specifications
About this item
Goat anti-Human IgA (alpha chain) - Affinity Pure, FITC Conjugate, min x w/bovine, mouse, and rabbit serum proteins
Type: Secondary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: α chain
Host: Goat
Isotype:
Reactivity: Human