Safety blood lancets, VITREX® STERILANCE® Lite III
Supplier: Vitrex Medical A/S
New Product
Certificates
About this item
Safety lancets for blood glucose and other in vitro diagnostic tests.
- Needle fully shielded before and after use
- Pre-loaded pressure activated design
- High quality, ultra sharp tri-bevel needle, high speed penetration
A variety of point configurations and penetration depths suit most capillary blood requirements.