ESD safety rigger boots, Safety-Grip, pull-on
Supplier: ELTEN
Certificates
About this item
These ESD winter safety boots are made from cowhide leather with a PU/PU walking sole (Safety-Grip).
- Warm lining
- Full length warm fleece, ESD inlay sole
- PU scuff cap protects the leather upper
- Metal-free, puncture resistant midsole
- Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Brown
Certifications: EN 61340-5-1 (ESD); EN ISO 20345; S3, SRC, CI.
Specifications
- Catalog No:
- 114-8232
- 114-8233
- 114-8235
- 114-8230
- 114-8231
- 114-8229
- 114-8227
- 114-8228
- 114-8238
- 114-8226
- 114-8239
- 114-8234
- 114-8236
- 114-8237
- 114-8225
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001