Test kits, photometric, Organic acids, LCK cuvette tests
Catalog # 705-3000
Supplier: Hach
CAS Number:
Specifications
- Chemical method:Esterification
- Pk:25 Tests
- Description:Organic acids
- Seq:259
- Type:LCK365
- For unit:DR3900, DR6000, DR1900, DR2800, DR3800, DR5000
- Range:50 - 2500 mg/l (CH₃COOH)
75 - 3600 mg/l (C₃H7COOH)
Specifications
About this item
Pre-dosed reagents for maximum safety.
- Precise and reliable measurement values
- Maximum safety for users, thanks to the closed system and low amounts of reagents
- Convenient and error-free dosing of the reagents
- Barcode label for automatic recognition in the photometer
- Differentiation between tests and measuring ranges by means of colour-coding
Storage conditions: 15 to 25 °C