Test kits, photometric, Nitrate, LCK cuvette tests
Supplier: Hach
Certificates
About this item
Pre-dosed reagents for maximum safety.
- Precise and reliable measurement values
- Maximum safety for users, thanks to the closed system and low amounts of reagents
- Convenient and error-free dosing of the reagents
- Barcode label for automatic recognition in the photometer
- Differentiation between tests and measuring ranges by means of colour-coding
Storage conditions: 15 to 25 °C
Certifications: ISO 7890-1-2-1986, DIN 38405 D9-2.
Related Information
Certificates
Search by Lot NumberHach LCK Cuvette Tests
Product Literature
HACHLCK339_TDS_EN