Order Entry
Norway
ContactUsLinkComponent
Sodium sulphate, anhydrous ≥99.0% (dried basis), powder, GenAR® USP for biotechnology, Macron Fine Chemicals™
Sodium sulphate, anhydrous ≥99.0% (dried basis), powder, GenAR® USP for biotechnology, Macron Fine Chemicals™
Certificates
undefined
Sodium sulphate, anhydrous ≥99.0% (dried basis), powder, GenAR® USP for biotechnology, Macron Fine Chemicals™
Certificates

Specification Test Results

  • Bulk Pharmaceutical Chemical:
    = 99.0 %
  • USP - Assay (Na₂SO₄) (dried basis):
    ≥99.0 %
  • USP - Acidity or Alkalinity:
    Passes Test
  • USP - Chloride (Cl):
    ≤0.005 %
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Identification C:
    Passes Test
  • USP - Insoluble Matter:
    ≤0.01 %
  • USP - Iron (Fe):
    ≤0.001 %
  • USP - Loss on Drying at 105°C:
    ≤0.5 %
  • Thru U.S. No. 50 Sieve, For Information Only:
    For Information Only
  • Solubility (5 in 100):
    Passes Test
  • EP/BP - Assay (Na₂SO₄):
    98.5 - 101.0 %
  • EP/BP - Acidity or Alkalinity:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Chloride (Cl):
    ≤450 ppm
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Identification C:
    Passes Test
  • EP/BP - Loss on Drying @ 130° C:
    ≤0.5 %
  • No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.:
    = 0.5 %

Specification Test Results

Related Information

Certificates
Search by Lot Number