Order Entry
Norway
ContactUsLinkComponent
 
Sodium sulphate, anhydrous ≥99.0%, granular, AR® ACS 10 - 60 Mesh, Macron Fine Chemicals™
 
undefined
Sodium sulphate, anhydrous ≥99.0%, granular, AR® ACS 10 - 60 Mesh, Macron Fine Chemicals™
 

 

  • Meets ACS Reagent Chemical Requirements:
    Passes Test
  • For Laboratory,Research,or Manufacturing Use:
    Min. 99.0 %
  • ACS - Appearance (white, crystalline granules):
    Passes Test
  • ACS - Assay (Na₂SO₄):
    ≥99.0 %
  • ACS - Calcium (Ca):
    ≤0.01 %
  • ACS - Chloride (Cl):
    ≤0.001 %
  • ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • ACS - Insoluble Matter:
    ≤0.01 %
  • ACS - Iron (Fe):
    ≤0.001 %
  • ACS - Magnesium (Mg):
    ≤0.005 %
  • ACS - Loss on Ignition:
    ≤0.5 %
  • ACS - Nitrogen Compounds (as N):
    ≤5 ppm
  • ACS - pH of 5% Solution at 25°C:
    5.2 - 9.2
  • ACS - Phosphate (PO₄):
    ≤0.001 %
  • ACS - Potassium (K):
    ≤0.01 %
  • Solubility and Foreign Matter:
    Passes Test
  • On U.S. No. 10 Sieve:
    ≤1 %
  • On U.S. No. 60 Sieve:
    ≥80 %
  • Thru U.S. No. 60 Sieve:
    ≤19 %
  • Thru U.S. No. 100 Sieve:
    ≤10 %
  • Extraction - Concentration Suitability:
    Passes Test