Order Entry
Norway
ContactUsLinkComponent
Sodium hydroxide 10 N in aqueous solution, Biotech Reagent, J.T.Baker®
Corrosive
Irritant
Sodium hydroxide 10 N in aqueous solution, Biotech Reagent, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Corrosive
Irritant
Sodium hydroxide 10 N in aqueous solution, Biotech Reagent, J.T.Baker®
Supplier:  Avantor
Certificates

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specification Test Results

  • GMP Manufactured Product:
    Passes Test
  • IMPORTANT: Material will freeze if stored below 41 °F (5°C).:
    9.9 - 10.1
  • Protect from air to avoid absorption of carbon dioxide.:
    = 0.6 %
  • CAUTION: For Manufacturing, processing or repackaging:
    = 5 ppm
  • Appearance (Clear, colorless liquid):
    Passes Test
  • Normality:
    9.9 - 10.1
  • Sodium Carbonate (Na₂CO₃):
    ≤0.6 %
  • Chloride (Cl):
    ≤20 ppm
  • ACS - Heavy Metals (as Pb):
    ≤1 ppm
  • Trace Impurities - Iron (Fe):
    ≤2 ppm
  • Endotoxin Concentration (EU/mL):
    ≤2.0

Specification Test Results

Related Information

Frequently Bought Together