Order Entry
Norway
ContactUsLinkComponent
Sodium hydrogen carbonate 99.0-100.5% (dried basis), powder USP, ACS, J.T.Baker®
Sodium hydrogen carbonate 99.0-100.5% (dried basis), powder USP, ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Sodium hydrogen carbonate 99.0-100.5% (dried basis), powder USP, ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Bulk Pharmaceutical Chemical:
    = 5 ppm
  • ACS - Ammonium (NH₄):
    ≤5 ppm
  • ACS - Assay (NaHCO₃) (by acidimetry):
    99.7 - 100.3 %
  • ACS - Calcium (Ca):
    ≤0.02 %
  • ACS - Chloride (Cl):
    ≤0.003 %
  • ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • ACS - Insoluble Matter:
    ≤0.015 %
  • ACS - Iron (Fe):
    ≤0.001 %
  • ACS - Magnesium (Mg):
    ≤0.005 %
  • ACS - Phosphate (PO₄):
    ≤0.001 %
  • ACS - Potassium (K):
    ≤0.005 %
  • ACS - Sulfur Compounds (as SO₄):
    ≤0.003 %
  • USP - Assay (NaHCO₃) (dried basis):
    99.0 - 100.5 %
  • USP - Chloride (Cl):
    ≤0.015 %
  • USP - Identification:
    Passes Test
  • USP - Insoluble Substances:
    Passes Test
  • USP - Limit of Ammonia:
    ≤20 ppm
  • USP - Limit of Sulfur Compounds:
    ≤0.015 %
  • USP - Loss on Drying:
    ≤0.25 %
  • USP - Normal Carbonate:
    Passes Test
  • USP - Arsenic (As):
    ≤2 ppm

Specification Test Results

Related Information

Certificates
Search by Lot Number