
Guanidiniumklorid ≥99.5%, NF-GenAR® for biotechnology, Macron Fine Chemicals™
:50-01-1
:
:NH₂C(=NH)NH₂·HCl
:MFCD00013026
:95.53 g/mol
:13,04578
:185 °C
:1,354 g/cm³ (20 °C)
:200-002-3
:GHS07
:Romtemperatur
- GMP Manufactured Product:≥99.5%
- CAUTION: For Manufacturing, processing or repackaging:≤0.01%
- Assay (NH₂C(:NH)NH₂·HCl):≥99.5 %
- Acidity (as HCl):≤0.01 %
- Arsenic and Antimony (as As):≤10 ppm
- Copper (Cu):≤10 ppm
- Heavy Metals (as Pb):≤10 ppm
- Iron (Fe):≤10 ppm
- Loss on Drying at 105°C:≤0.5 %
- Enzyme Activity - DNase Activity:Passes Test
- Enzyme Activity - RNase Activity:Passes Test
- Enzyme Activity - Protease Activity:Passes Test
- Melamine and Related Compounds:Passes Test
- Absorbance of a 6M Solution (260 nm) (au):≤0.030
- Residue after Ignition:≤0.20 %
- Water-Insoluble Matter:≤0.3 %
Frequently Bought Together