Order Entry
Norway
ContactUsLinkComponent
Dichloromethane stabilised, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®
Irritant
Health
Dichloromethane stabilised, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Irritant
Health
Dichloromethane stabilised, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay (by GC):
    min. 99.8 %
  • Acidity:
    max. 0.0005 meq/g
  • Evaporation Residue:
    max. 5 ppm
  • GC sensitive impurities (lindane GC-ECD):
    max. 5 ng/l
  • Stabiliser (Amylene):
    about 50 ppm
  • Water (KF):
    max. 0.05 %

Specification Test Results

Related Information