Order Entry
Norway
ContactUsLinkComponent
Organic reference standards, estradiols, Ph. Eur.
Organic reference standards, estradiols, Ph. Eur.
Supplier:  EDQM
Certificates
Organic reference standards, estradiols, Ph. Eur.
Supplier:  EDQM

Specifications

  • CAS No.:
    35380-71-3
  • Formula:
    C₁₈H₂₄O₂·₀.₅H₂O
  • Description:
    Estradiol hemihydrate

Specifications