Order Entry
Norway
ContactUsLinkComponent
Organic reference standards, desogestrels, Ph. Eur.
Organic reference standards, desogestrels, Ph. Eur.
Supplier:  EDQM
Certificates
Organic reference standards, desogestrels, Ph. Eur.
Supplier:  EDQM

Specifications

  • CAS No.:
    54024-22-5
  • Formula:
    C₂₂H₃₀O
  • Description:
    Desogestrel

Specifications