kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
Standards
Organic Standards
Organic reference standards, clotrimazoles, Ph. Eur.
Organic reference standards, clotrimazoles, Ph. Eur.
:
EDQM
Organic reference standards, clotrimazoles, Ph. Eur.
:
EDQM
CAS No.:
23593-75-1
Formula:
C₂₂H₁₇ClN₂
Description:
Clotrimazole for peak identification, Ph. Eur. standard