Transport boxes, VACUETTE®
Catalog # GREI472001_10
Supplier: Greiner Bio-One
Specifications
- Description:Transport box Vacuette
- Pk:10
Specifications
About this item
Transport box made of PP.
- For transport of up to 40 tubes of diameter 9 to 16 mm
- With practical snap closure
- Leak-proof, due to sealing ring and absorbent pad
- Stackable
- Separate slot for cooler pack
Ensure the safe transport of sample material from the collection point right up to analysis in the laboratory.