Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Organic reference standards, brompheniramine maleate, Ph. Eur.
Organic reference standards, brompheniramine maleate, Ph. Eur.
EDQMB1153000
:
EDQM
:
Organic reference standards, brompheniramine maleate, Ph. Eur.
EDQMB1153000
:
EDQM
:
Pk:
100 mg
Standards:
Reference standard
CAS No.:
980-71-2
Description:
Brompheniramine maleate
Formula:
C₂₀H₂₃BrN₂O₄
Description:
Brompheniramine maleate