kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
Standards
Organic Standards
Organic reference standards, betamethasone dipropionate, Ph. Eur.
Organic reference standards, betamethasone dipropionate, Ph. Eur.
:
EDQM
Organic reference standards, betamethasone dipropionate, Ph. Eur.
:
EDQM
CAS No.:
5593-20-4
Formula:
C₂₈H₃₇FO₇
Description:
Betamethasone dipropionate for peak identification