Anti-HIST1H2BB Rabbit Monoclonal Antibody [clone: RM235]
ABNOMAB15112
: Abnova
- Antibody type:Primary
- Antigen symbol:HIST1H2BB
- Clonality:Monoclonal
- Clone:RM235
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:3018
- Storage buffer:In PBS (50% glycerol, 1% BSA, 0.09% sodium azide)
- Storage temperature:Store at -20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic acetyl peptide corresponding to residues surrounding K20 of human histone H2B.
- Purification:Protein A affinity purification
- Size:100 µg
- Pk:100 µG
Rabbit monoclonal antibody raised against synthetic acetyl peptide of human histone H2B (acetyl K20).
Type: Primary
Antigen: HIST1H2BB
Clonality: Monoclonal
Clone: RM235
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human