Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Electrochemistry Meters
pH and Multi-Parameter Meters
Accessories for Portamess 911 / 913
Accessories for Portamess 911 / 913
Accessories for Portamess 911 / 913
Supplier:
KNICK
Certificates
Expand image
Accessories for Portamess 911 / 913
Supplier:
KNICK
About this item
Specifications
Catalog No:
663-2215
662-3537
662-3517
662-3516
662-3539
662-3532
Model:
ZU 6959
pH range:
-
Temperature range:
–10...+100 °C
Specifications