Anti-RGCC Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BOSSBS-9079R-HRP
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Regulator of Cell Cycle
- Antigen symbol:RGCC
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Gene ID:28984
- Antigen synonyms:C13orf15|regulator of cell cycle|bA157L14.2|RGC32|RGC-32
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:98-137/137
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Modulates the activity of cell cycle-specific kinases. Enhances CDK1 activity. May contribute to the regulation of the cell cycle. May inhibit growth of glioma cells by promoting arrest of mitotic progression at the G2/M transition. Fibrogenic factor contributing to the pathogenesis of renal fibrosis through fibroblast activation.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: RGCC
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat