Order Entry
Norway
ContactUsLinkComponent
Anti-NOL4L Rabbit Polyclonal Antibody (Cy3®)
Anti-NOL4L Rabbit Polyclonal Antibody (Cy3®)
  BOSSBS-9693R-CY3
 :  Bioss
Anti-NOL4L Rabbit Polyclonal Antibody (Cy3®)
  BOSSBS-9693R-CY3
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Nucleolar Protein 4 Like
  • Antigen symbol:
    NOL4L
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy3®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    140688
  • Antigen synonyms:
    dJ1184F4.2|C20orf112|dJ1184F4.4|C20orf113
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

Representing about 2% of human DNA, chromosome 20 consists of approximately 63 million bases and 600 genes. Chromosome 20 contains a region with numerous genes expressed in the epididymis, which are thought important for seminal production, and some viewed as potential targets for male contraception. The PRNP gene encoding the prion protein associated with spongiform encephalopathies, like Creutzfeldt-Jakob disease, is found on chromosome 20. Amyotrophic lateral sclerosis, spinal muscular atrophy, ring chromosome 20 epilepsy syndrome and Alagille syndrome are also associated with chromosome 20. The C20orf112 gene product has been provisionally designated C20orf112 pending further characterization.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NOL4L
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat