Anti-NUPR1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
BOSSBS-7106R-A350
: Bioss
- Antibody type:Primary
- Antigen name:Nuclear Protein, Transcriptional Regulator, 1
- Antigen symbol:NUPR1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:26471
- Antigen synonyms:Nuclear protein 1|Candidate of metastasis 1|NUPR1|NUPR1_HUMAN|p8 protein|Protein p8.|COM1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Could participate in the response to proapoptotic stimuli and promotes cellular growth in a way that helps the tissue counteract diverse injuries. May contribute to the metastatic phenotype.Tissue specificity: Highly expressed in pancreas.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NUPR1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat