Anti-CCDC47 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-6922R-CY5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:coiled-coil Domain Containing 47
- Antigen symbol:CCDC47
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rabbit,Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:57003
- Antigen synonyms:GK001|MSTP041
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
CCDC47
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CCDC47
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Rabbit