Anti-MAGEA12 Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-6818R-CY5.5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Melanoma Antigen Family A12
- Antigen symbol:MAGEA12
- Clonality:Polyclonal
- Conjugation:Cy5.5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Gene ID:4111
- Antigen synonyms:CT1.12|MAGE12
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Not known, though may play a role in tumor transformation or aspects of tumor progression.Tissue specificity: Expressed in many tumors of several types, such as melanoma, head and neck squamous cell carcinoma, lung carcinoma and breast carcinoma, but not in normal tissues except for testes.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MAGEA12
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human