Anti-FOLH1B Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
BOSSBS-6535R-FITC
: Bioss
- Antibody type:Primary
- Antigen name:Folate Hydrolase 1B
- Antigen symbol:FOLH1B
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:PSM|GCPIII|PSMA-LIKE|GCP3|PSMAL|FOLHP|FOLH2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
FOLH1B
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: FOLH1B
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat