Anti-CHORDC1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
BOSSBS-6409R-A488
: Bioss
- Antibody type:Primary
- Antigen name:Cysteine And Histidine-Rich Domain Containing 1
- Antigen symbol:CHORDC1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rabbit,Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:26973
- Antigen synonyms:CHP1|Cysteine and histidine-rich domain-containing protein 1|CHORD-containing protein 1|CHORD domain-containing protein 1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
CHORDC1
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CHORDC1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity: