Order Entry
Norway
ContactUsLinkComponent
Anti-TMPRSS4 Rabbit Polyclonal Antibody (Cy5®)
Anti-TMPRSS4 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-6286R-CY5
Supplier:  Bioss
Anti-TMPRSS4 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-6286R-CY5
Supplier:  Bioss

Specifications

  • Antistofftype:
    Primary
  • Antigen-navn:
    Transmembrane Protease Serine 4
  • Antigensymbol:
    TMPRSS4
  • Klonalitet:
    Polyclonal
  • Konjugering:
    Cy5®
  • Flowcytometri:
    Yes
  • Vert:
    Rabbit
  • ImmunoKjemi:
    Yes
  • Isotype:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Kyssabsorpsjon:
    No
  • Skjema:
    liquid
  • Gen-ID:
    56649
  • Antigen synonymer:
    TMPRSS3|transmembrane serine protease 3|membrane-type serine protease 2|serine 4|MT-SP2|transmembrane protease|type II membrane serine protease
  • Lagringsbuffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Lagringstemp:
    Store at 4°C for 12 months
  • Konsentrasjon:
    1 μg/μl
  • Frakttemp.:
    4°C
  • Rensing:
    Purified by Protein A
  • Fp:
    100 µl

Specifications

About this item

TMPRSS4 is a member of the peptidase S1 family and contains 1 LDL receptor class A domain, 1 peptidase S1 domain and 1 SRCR domain. It is a probable membrane protease capable of activating ENaC and may process sodium channels in endothelial cells. TMPRSS4 is overexpressed in thyroid neoplasms, and splice variants in TMPRSS4 are thought to be linked with different cancers. Three named isoforms are produced by alternative splicing.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: TMPRSS4
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat