Anti-RAB20 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
BOSSBS-6178R-A488
: Bioss
- Antibody type:Primary
- Antigen name:Ras-related protein Rab-20
- Antigen symbol:RAB20
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:55647
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:150-180/234
- Purification:Purified by Protein A
- Pk:100 µl
Plays a role in apical endocytosis/recycling. Plays a role in the maturation and acidification of phagosomes that engulf pathogens, such as S.aureus and M.tuberculosis. Plays a role in the fusion of phagosomes with lysosomes.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RAB20
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat