Anti-WNT8A Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-6129R-A350
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Wingless-type MMTV Integration Site Family, Member 8A
- Antigen symbol:WNT8A
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:7478
- Antigen synonyms:Stra11|Wnt-8A|Wnt-8D|Wnt8d
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:200-250/351
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Ligand for members of the frizzled family of seven transmembrane receptors. May play an important role in the development and differentiation of certain forebrain structures, notably the hippocampus.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: WNT8A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat