Anti-SHPRH Rabbit Polyclonal Antibody (Alexa Fluor® 350)
BOSSBS-6104R-A350
: Bioss
- Antibody type:Primary
- Antigen name:SNF2 histone linker PHD RING helicase
- Antigen symbol:SHPRH
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:257218
- Antigen synonyms:E3 ubiquitin-protein ligase SHPRH|SNF2 histone linker PHD RING helicase|PHD and RING finger domain-containing helicase|SNF2|SHPRH_HUMAN.|histone-linker
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
SHPRH is a ubiquitously expressed protein that contains motifs characteristics of several DNA repair proteins, transcription factors, and helicases. SHPRH is a functional homolog of S. cerevisiae RAD5
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SHPRH
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat