Order Entry
Norway
ContactUsLinkComponent
Anti-RASAL Rabbit Polyclonal Antibody (Cy5.5®)
Anti-RASAL Rabbit Polyclonal Antibody (Cy5.5®)
Katalog # BOSSBS-6088R-CY5.5
Supplier:  Bioss
Anti-RASAL Rabbit Polyclonal Antibody (Cy5.5®)
Katalog # BOSSBS-6088R-CY5.5
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    RAS Protein Activator Like 1
  • Antigen symbol:
    RASAL
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5.5®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    8437
  • Antigen synonyms:
    RAS protein activator like 1 (GAP1 like)|GAP1 like protein|RASAL|RAS-GTPase-activating protein-like|RAS protein activator like 1|RASAL1
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    460-510/805
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Probable inhibitory regulator of the Ras-cyclic AMP pathway.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RASAL
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat