Anti-NOV Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-6059R-CY3
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Nephroblastoma overexpressed
- Antigen symbol:NOV
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:4856
- Antigen synonyms:IBP-9|IGFBP-9|NOVh|IGFBP9|CCN3
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:250-300/357
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Immediate-early protein likely to play a role in cell growth regulation.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NOV
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat