Specifications
- Antibody type:Primary
- Antigen name:SET And MYND Domain Containing 3
- Antigen symbol:SMYD3
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:64754
- Antigen synonyms:Zinc finger MYND domain-containing protein 1|SMYD3|ZMYND1|ZNFN3A1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
SMYD3 is a gene that is over-expressed in the majority of colorectal carcinomas and hepatocellular carcinomas. SMYD3 forms a complex with RNA polymerase II through an interaction with the RNA helicase HELZ and transactivates a set of genes that included oncogenes, homeobox genes and genes associated with cell-cycle regulation. SMYD3 binds to a motif, 5'-CCCTCC-3', present in the promoter region of downstream genes such as Nkx2.8. The SET domain of SMYD3 shows histone H3-lysine 4 (H3-K4)-specific methyltransferase activity, which is enhanced in the presence of the heat-shock protein HSP90A.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SMYD3
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat