Order Entry
Norway
ContactUsLinkComponent
Anti-HEPACAM Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anti-HEPACAM Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-5840R-A488
Supplier:  Bioss
Anti-HEPACAM Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-5840R-A488
Supplier:  Bioss

Specifications

  • Antistofftype:
    Primary
  • Antigen-navn:
    hepatic and glial cell adhesion molecule
  • Antigensymbol:
    HEPACAM
  • Klonalitet:
    Polyclonal
  • Konjugering:
    Alexa Fluor® 488
  • Vert:
    Rabbit
  • ImmunoKjemi:
    Yes
  • Isotype:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Kyssabsorpsjon:
    No
  • Skjema:
    liquid
  • Gen-ID:
    220296
  • Antigen synonymer:
    MLC2B|MLC2A|GlialCAM
  • Lagringsbuffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Lagringstemp:
    Store at 4°C for 12 months
  • Konsentrasjon:
    1 μg/μl
  • Frakttemp.:
    4°C
  • Rensing:
    Purified by Protein A
  • Fp:
    100 µl

Specifications

About this item

Involved in regulating cell motility and cell-matrix interactions. May inhibit cell growth through suppression of cell proliferation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HEPACAM
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat