Anti-CABLES2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
BOSSBS-5744R-A488
: Bioss
- Antibody type:Primary
- Antigen name:Cdk5 And Abl enzyme substrate 2
- Antigen symbol:CABLES2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:81928
- Antigen synonyms:Ik3 2 |C20orf150|Interactor with CDK3 2|CDK5 and ABL1 enzyme substrate 2|CABL2_HUMAN.
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
CABLES2 is a proapoptotic factor involved in both p53-mediated and p53-independent apoptotic pathways. ik3-1/CABLES1, a closely related molecule, has been identified to be a candidate tumor suppressor for colon and head/neck cancers.The exact function of this protein is unknown, but it is probably involved in G1-S cell cycle transition. CABLES2 binds to CDK3, CDK5 and ABL1.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CABLES2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat