Anti-PIK3C3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
BOSSBS-5580R-HRP
: Bioss
- Antibody type:Primary
- Antigen name:Phosphatidylinositol-4-Phosphate 3-Kinase, Catalytic Subunit Type 3
- Antigen symbol:PIK3C3
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Gene ID:5289
- Antigen synonyms:Phosphatidylinositol 3-kinase catalytic subunit type 3|Phosphatidylinositol 3-kinase p100 subunit|PI3-kinase type 3|PIK3C3|hVps34|VPS34
- Modification:phospho-Ser676
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
PI 3 Kinase Class 3 is a member of the PI3/PI4-kinase family. It is the catalytic subunit of the PI3K complex. It is involved in the transport of lysosomal enzyme precursors to lysosomes. It is ubiquitously expressed, with a highest expression in skeletal muscle.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: PIK3C3
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat