Order Entry
Norway
ContactUsLinkComponent
Anti-MEF2D Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-MEF2D Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Katalog # BOSSBS-5501R-FITC
Leverandør:  Bioss
Anti-MEF2D Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Katalog # BOSSBS-5501R-FITC
Leverandør:  Bioss

Spesifikasjoner

  • Antistofftype:
    Primary
  • Antigen-navn:
    Myocyte Enhancer Factor 2D
  • Antigensymbol:
    MEF2D
  • Klonalitet:
    Polyclonal
  • Konjugering:
    FITC (Fluorescein Isothiocyanate)
  • Vert:
    Rabbit
  • ImmunoKjemi:
    Yes
  • Isotype:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Environmentally Preferable:
  • Kyssabsorpsjon:
    No
  • Skjema:
    liquid
  • Gen-ID:
    4209
  • Antigen synonymer:
    polypeptide D (myocyte enhancer factor 2D)|myocyte enhancer factor 2D/deleted in azoospermia associated protein 1 fusion protein|myocyte-specific enhancer factor 2D|MADS box transcription enhancer factor 2|MEF2D/DAZAP1 fusion
  • Modifikasjon:
    phospho-Ser180
  • Lagringsbuffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Lagringstemp:
    Store at 4°C for 12 months
  • Konsentrasjon:
    1 μg/μl
  • Frakttemp.:
    4°C
  • Rensing:
    Purified by Protein A
  • Fp:
    100 µl

Spesifikasjoner

Om dette produktet

Transcriptional activator which binds specifically to the MEF2 element, 5'-YTA[AT](4)TAR-3', found in numerous muscle-specific, growth factor- and stress-induced genes. Mediates cellular functions not only in skeletal and cardiac muscle development, but also in neuronal differentiation and survival. Plays diverse roles in the control of cell growth, survival and apoptosis via p38 MAPK signaling in muscle-specific and/or growth factor-related transcription. Plays a critical role in the regulation of neuronal apoptosis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MEF2D
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat