Anti-PHYH Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-5091R-A555
Leverandør: Bioss
Spesifikasjoner
- Antibody type:Primary
- Antigen name:Phytanoyl-CoA 2-hydroxylase
- Antigen symbol:PHYH
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:5264
- Antigen synonyms:Phytanoyl-CoA dioxygenase peroxisomal|Phytanic acid oxidase|Phytanoyl-CoA alpha-hydroxylase|PAHX|PHYH
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:40-90/338
- Purification:Purified by Protein A
- Pk:100 µl
Spesifikasjoner
Om dette produktet
Converts phytanoyl-CoA to 2-hydroxyphytanoyl-CoA.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PHYH
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat