Spesifikasjoner
- Antibody type:Primary
- Antigen name:Chromosome 7 open reading frame 65
- Antigen symbol:C7ORF65
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Gene ID:401335
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Spesifikasjoner
Om dette produktet
Endocannaboids such as arachidonoyl ethanolamide and 2-arachidonoyl glycerol function as short range modulators of cell and synaptic activity. Monoacylglycerol lipase hydrolyzes 2-arachidonoyl glycerol which terminates it's biological actions. Monoacylglycerol lipase also works consecutively with hormone sensitive lipase to mobilize fatty acids from triglyceride stores of adipocytes. Monoacylglycerol lipase is expressed in kidney, spleen, heart, liver, testis, stomach, brain and lung tissue, and adrenal gland. It is most abundantly expressed in skeletal muscle and adipose tissue.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: C7ORF65
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat