Anti-PFKFB1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-5004R-A555
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 1
- Antigen symbol:PFKFB1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:5207
- Antigen synonyms:6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 1|F6PK|6PF-2-K/Fru-2,6-P2ASE liver isozyme
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
PFKFB1 belongs to the family of bifunctional 6-phosphofructo-2-kinase:fructose-2,6-biphosphatase enzymes. It forms a homodimer that is involved in both the synthesis and degradation of fructose 2,6-bisphosphate. The activity of this enzyme is thought to regulate glucose homeostasis.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PFKFB1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat