Anti-CDC40 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-4295R-CY7
Leverandør: Bioss
Spesifikasjoner
- Antibody type:Primary
- Antigen name:Cell Division Cycle 40
- Antigen symbol:CDC40
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:51362
- Antigen synonyms:EH-binding protein 3|Pre-mRNA-processing factor 17|Ehb3|Cell Division Cycle 40 homolog|PRP17 homolog|PRPF17,|hPRP17|PRP17|CDC40|EHB3
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:515-565/579
- Purification:Purified by Protein A
- Pk:100 µl
Spesifikasjoner
Om dette produktet
Associates with the spliceosome late in the splicing pathway and may function in the second step of pre-mRNA splicing.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CDC40
Clonality: Polyclonal
Clone: 0
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: