Anti-ARHGEF12 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-4213R-A647
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Rho Guanine Nucleotide Exchange Factor (GEF) 12
- Antigen symbol:ARHGEF12
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:23365
- Antigen synonyms:PRO2792|LARG
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Rho GTPases play a fundamental role in numerous cellular processes that are initiated by extracellular stimuli that work through G protein coupled receptors. The encoded protein may form a complex with G proteins and stimulate Rho-dependent signals. This protein is observed to form myeloid/lymphoid fusion partner in acute myeloid leukemia (from EntrezGene). LARG has also been implicated in linking heterotrimeric G proteins to microtubule organisation in regulation of cell polarity.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ARHGEF12
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat