Anti-CD19 Rabbit Polyclonal Antibody (Cy5®)
BOSSBS-4755R-CY5
: Bioss
- Antibody type:Primary
- Antigen name:CD19 molecule
- Antigen symbol:CD19
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:930
- Antigen synonyms:B-lymphocyte antigen CD19. B4|CVID3|AW495831|mouse CD19|Cluster of Differentiation 19
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:171-220/556
- Purification:Purified by Protein A
- Pk:100 µl
Assembles with the antigen receptor of B-lymphocytes in order to decrease the threshold for antigen receptor-dependent stimulation.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CD19
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat