Order Entry
Norway
ContactUsLinkComponent
Anti-WFDC2 Rabbit Polyclonal Antibody (Cy5.5®)
Anti-WFDC2 Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-4626R-CY5.5
Supplier:  Bioss
Anti-WFDC2 Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-4626R-CY5.5
Supplier:  Bioss

Specifications

  • Antistofftype:
    Primary
  • Antigen-navn:
    WAP four-disulfide core Domain 2
  • Antigensymbol:
    WFDC2
  • Klonalitet:
    Polyclonal
  • Konjugering:
    Cy5.5®
  • Vert:
    Rabbit
  • ImmunoKjemi:
    Yes
  • Isotype:
    IgG
  • Reaktivitet:
    Rat,
    Mouse
  • Kyssabsorpsjon:
    No
  • Skjema:
    liquid
  • Gen-ID:
    67701
  • Antigen synonymer:
    epididymal protein 4|WFDC2|four-disulfide core|EDDM4|WAP domain containing protein HE4-V4|WAP four-disulfide core domain 2|WAP5|MGC57529|epididymal secretory protein E4|epididymis-specific|major epididy|whey-acidic protein type|dJ461P17.6|HE4
  • Lagringsbuffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Lagringstemp:
    Store at 4°C for 12 months
  • Konsentrasjon:
    1 μg/μl
  • Frakttemp.:
    4°C
  • Immunogen:
    110-160/174
  • Rensing:
    Purified by Protein A
  • Fp:
    100 µl

Specifications

About this item

Broad range protease inhibitor.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: WFDC2
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat