Anti-AURKAIP1 Rabbit Polyclonal Antibody (Cy5®)
BOSSBS-4287R-CY5
: Bioss
- Antibody type:Primary
- Antigen name:Aurora Kinase AIP1
- Antigen symbol:AURKAIP1
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:54998
- Antigen synonyms:AIP|MRP-S38|AKIP
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:150-199/199
- Purification:Purified by Protein A
- Pk:100 µl
May act as a negative regulator of Aurora-A kinase, by down-regulation through proteasome-dependent degradation.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: AURKAIP1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat